Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Arachidonylcyclopropylamide, 98%

Chemical Identifiers

Chemical Name or Material N-Arachidonylcyclopropylamide
CAS 229021-64-1
Formula Weight 343.55
MDL Number MFCD02683582
Synonym ACPA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67376MA

 
alfa aesar™
J67376MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 343.55
Chemical Name or Material N-Arachidonylcyclopropylamide
Assay 98%
CAS 229021-64-1
Sensitivity Air Sensitive
Physical Form Liquid
MDL Number MFCD02683582
Synonym ACPA
Color Pale yellow
Quantity 10mg
Storage Note 1 -30° to -10°C
RTECS Number JX3841600
Safety and Handling

-30° to -10°C