Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Arachidonyl-2-chloroethylamide, 98%


Quantity 10mg

Manufacturer: alfa aesar™  J67294MA

 View more versions of this product

Catalog No. AAJ67294MA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 365.99
Chemical Name or Material N-Arachidonyl-2-chloroethylamide
MDL Number MFCD02683581
CAS 220556-69-4
Assay 98%
Physical Form Oil
Synonym ACEA; N-(2-Chloroethyl)-5Z,8Z,11Z,14Z-eicosatetraenamide
Color Pale yellow
Quantity 10mg
Storage Note 1 -30° to -10°C
Sensitivity Air Sensitive
RTECS Number JX3840800

Specifications

Chemical Name or Material N-Arachidonyl-2-chloroethylamide
CAS 220556-69-4
Formula Weight 365.99
MDL Number MFCD02683581
Synonym ACEA; N-(2-Chloroethyl)-5Z,8Z,11Z,14Z-eicosatetraenamide
Accessories

-30° to -10°C