Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Arachidonyl-2-chloroethylamide, 98%

Chemical Identifiers

Chemical Name or Material N-Arachidonyl-2-chloroethylamide
CAS 220556-69-4
Formula Weight 365.99
MDL Number MFCD02683581
Synonym ACEA; N-(2-Chloroethyl)-5Z,8Z,11Z,14Z-eicosatetraenamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67294MA

 
alfa aesar™
J67294MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 365.99
Chemical Name or Material N-Arachidonyl-2-chloroethylamide
MDL Number MFCD02683581
CAS 220556-69-4
Assay 98%
Physical Form Oil
Synonym ACEA; N-(2-Chloroethyl)-5Z,8Z,11Z,14Z-eicosatetraenamide
Color Pale yellow
Quantity 10mg
Storage Note 1 -30° to -10°C
Sensitivity Air Sensitive
RTECS Number JX3840800
Safety and Handling

-30° to -10°C