Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Arachidonoyldopamine, 98%

Chemical Identifiers

Chemical Name or Material N-Arachidonoyldopamine
CAS 199875-69-9
Formula Weight 439.64
MDL Number MFCD03412031
Synonym N-Arachidonoyl-3-hydroxytyramine; NADA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67250LB0

 
alfa aesar™
J67250LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67250MB

 
alfa aesar™
J67250MB
25mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 439.64
Chemical Name or Material N-Arachidonoyldopamine
Assay 98%
MDL Number MFCD03412031
CAS 199875-69-9
Physical Form Viscous Oil
Synonym N-Arachidonoyl-3-hydroxytyramine; NADA
Storage Note 1 -30° to -10°C
Sensitivity Air Sensitive
Quantity 5mg
Safety and Handling

-30° to -10°C