Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Arachidonoyl-serotonin, 98%

Chemical Identifiers

Chemical Name or Material N-Arachidonoyl-serotonin
CAS 187947-37-1
Formula Weight 462.68
MDL Number MFCD02179189
Synonym N-AA-5HT
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67271MA

 
alfa aesar™
J67271MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 462.68
Chemical Name or Material N-Arachidonoyl-serotonin
Physical Form Oil
Assay 98%
MDL Number MFCD02179189
CAS 187947-37-1
Color Pale yellow
Sensitivity Air & Light Sensitive
Quantity 10mg
Synonym N-AA-5HT
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C