Learn More
n-Amylamine, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 173942500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
n-Amylamine | |
98.5 | |
0.7500g/mL | |
4°C | |
98.5% min. (GC) | |
C5H13N | |
CH3(CH2)4NH2 | |
250mL | |
0.75 | |
amylamine, 1-aminopentane, pentylamine, 1-pentylamine, n-amylamine, 1-pentanamine, n-pentylamine, monoamylamine, norleucamine, amyl amine | |
DPBLXKKOBLCELK-UHFFFAOYSA-N | |
pentan-1-amine | |
8060 | |
CHEBI:74848 | |
99% |
110-58-7 | |
100.0 | |
104.0°C | |
Authentic | |
Glass bottle | |
1.4100 to 1.4120 | |
MFCD00008236 | |
, 175 | |
15, 593 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and ether | |
CCCCCN | |
87.15 | |
1.2 mPa.s (20°C) | |
87.15 |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/sh
GHS Signal Word: Danger
EINECSNumber : 203-780-2
Recommended Storage : Product may darken during storage
RUO â Research Use Only