Learn More
n-Amyl acetate, 99%, Thermo Scientific Chemicals
n-Amyl acetate, 99%, C7H14O2, CAS Number-628-63-7, 1-pentyl acetate, acetic acid, pentyl ester, amyl acetate, amyl acetic ester, amyl acetic ether, birnenoel, n-amyl acetate, n-pentyl acetate, n-pentyl ethanoate, pent-acetate, 2.5L, 98.5% min. (GC), 02, 131, 149.0 deg.C, 100.0, CHEBI:87362
Supplier: Thermo Scientific Chemicals 149180010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
n-Amyl acetate | |
98.5 | |
-70.8°C | |
0.8700g/mL | |
24°C | |
98.5% min. (GC) | |
C7H14O2 | |
CH3CO2(CH2)4CH3 | |
1 L | |
0.87 | |
Solubility in water: 10g/L (20c). Other solubilities: miscible with alcohol and ether | |
CCCCCOC(=O)C | |
130.19 | |
CHEBI:87362 | |
99% |
628-63-7 | |
100.0 | |
Colorless | |
149.0°C | |
Authentic | |
Glass bottle | |
1.4010 to 1.4030 | |
MFCD00009500 | |
02, 131 | |
amyl acetate, n-amyl acetate, acetic acid, pentyl ester, n-pentyl acetate, amyl acetic ester, 1-pentyl acetate, n-pentyl ethanoate, birnenoel, pent-acetate, amyl acetic ether | |
PGMYKACGEOXYJE-UHFFFAOYSA-N | |
pentyl acetate | |
12348 | |
130.19 | |
Liquid |
Chemical Identifiers
628-63-7 | |
130.19 | |
PGMYKACGEOXYJE-UHFFFAOYSA-N | |
12348 | |
pentyl acetate |
C7H14O2 | |
MFCD00009500 | |
amyl acetate, n-amyl acetate, acetic acid, pentyl ester, n-pentyl acetate, amyl acetic ester, 1-pentyl acetate, n-pentyl ethanoate, birnenoel, pent-acetate, amyl acetic ether | |
CHEBI:87362 | |
CCCCCOC(=O)C |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Warning
EINECSNumber : 211-047-3
RUO – Research Use Only