missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Amidinothiourea, 98+%

$54.16 - $636.45

Chemical Identifiers

CAS 2114-02-5
Molecular Formula C2H6N4S
Molecular Weight (g/mol) 118.16
MDL Number MFCD00014472
InChI Key OKGXJRGLYVRVNE-UHFFFAOYSA-N
Synonym 2-imino-4-thiobiuret, amidinothiourea, diaminomethylidene thiourea, guanylthiourea, guthimin, guthimine, gutimine, n-amidinothiourea, thiodicyanodiamidine, thiourea, aminoiminomethyl
PubChem CID 2724563
IUPAC Name diaminomethylidenethiourea
SMILES C(=NC(=S)N)(N)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1510114
View Documents
Alfa Aesar™
A1510114
25g Each for $54.16
 
AAA1510122
View Documents
Alfa Aesar™
A1510122
100g Each for $157.99
 
AAA1510136
View Documents
Alfa Aesar™
A1510136
500g Each for $636.45
 
Description

Description

Applications
2-Imino-4-thiobiuret is used in the design and synthesis of guanylthiourea derivatives which are potential inhibitors of plasmodium falciparum dihydrofolate reductase enzyme. 2-Imino-4-thiobiuret is also used as a reagent in recyclization reactions of N-arylmaleimides with polynucleophilic compounds.

Solubility
Soluble in water (70g/L).

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Chemical Identifiers

2114-02-5
118.16
OKGXJRGLYVRVNE-UHFFFAOYSA-N
2724563
C(=NC(=S)N)(N)N
C2H6N4S
MFCD00014472
2-imino-4-thiobiuret, amidinothiourea, diaminomethylidene thiourea, guanylthiourea, guthimin, guthimine, gutimine, n-amidinothiourea, thiodicyanodiamidine, thiourea, aminoiminomethyl
diaminomethylidenethiourea

Specifications

Odorless
25g
C2H6N4S
1756620
Soluble in water (70g/L).
NC(N)=NC(N)=S
118.16
118.16
N-Amidinothiourea
160°C to 168°C
2114-02-5
MFCD00014472
+--4-pentyn-2-ol, +/--4-pentyn-2-ol, 1-methyl-3-butyn-1-ol, 4-hydroxypentyne, 4-pentin-2-ol, 4-pentyn-2-ol, 4-pentyne-2-ol, pentyne-4-ol, r,s-4-pentyn-2-ol
OKGXJRGLYVRVNE-UHFFFAOYSA-N
(diaminomethylidene)thiourea
92915
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H361-H302
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319

EINECSNumber : 218-308-0

RTECSNumber : YR8250000

TSCA : No

Recommended Storage : Ambient temperatures