missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Amidinothiourea, 98+%

Manufacturer:  Alfa Aesar™ A1510136

 View more versions of this product

Catalog No. AAA1510136



Description

Description

Applications
2-Imino-4-thiobiuret is used in the design and synthesis of guanylthiourea derivatives which are potential inhibitors of plasmodium falciparum dihydrofolate reductase enzyme. 2-Imino-4-thiobiuret is also used as a reagent in recyclization reactions of N-arylmaleimides with polynucleophilic compounds.

Solubility
Soluble in water (70g/L).

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Specifications

N-Amidinothiourea
160°C to 168°C
2114-02-5
MFCD00014472
amidinothiourea, guanylthiourea, guthimin, guthimine, 2-imino-4-thiobiuret, n-amidinothiourea, thiourea, aminoiminomethyl, thiodicyanodiamidine, diaminomethylidene thiourea, gutimine
C(=NC(=S)N)(N)N
118.16
118.16
Odorless
500g
C2H6N4S
1756620
OKGXJRGLYVRVNE-UHFFFAOYSA-N
diaminomethylidenethiourea
2724563
≥98%

Chemical Identifiers

2114-02-5
118.16
OKGXJRGLYVRVNE-UHFFFAOYSA-N
2724563
C(=NC(=S)N)(N)N
C2H6N4S
MFCD00014472
amidinothiourea, guanylthiourea, guthimin, guthimine, 2-imino-4-thiobiuret, n-amidinothiourea, thiourea, aminoiminomethyl, thiodicyanodiamidine, diaminomethylidene thiourea, gutimine
diaminomethylidenethiourea
Safety and Handling

Safety and Handling

GHS H Statement
H361-H302
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.

GHS P Statement
P280h-P330-P501a
Wear protective gloves/clothing.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : YR8250000

Documents