missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Amidinothiourea, 98+%

Manufacturer:  Alfa Aesar™ A1510114

 View more versions of this product

Catalog No. AAA1510114



Description

Description

Applications
2-Imino-4-thiobiuret is used in the design and synthesis of guanylthiourea derivatives which are potential inhibitors of plasmodium falciparum dihydrofolate reductase enzyme. 2-Imino-4-thiobiuret is also used as a reagent in recyclization reactions of N-arylmaleimides with polynucleophilic compounds.

Solubility
Soluble in water (70g/L).

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Specifications

N-Amidinothiourea
160°C to 168°C
2117-11-5
MFCD00014472
4-pentyn-2-ol, 4-pentyne-2-ol, +--4-pentyn-2-ol, r,s-4-pentyn-2-ol, 4-hydroxypentyne, pentyne-4-ol, 4-pentin-2-ol, 1-methyl-3-butyn-1-ol, +/--4-pentyn-2-ol, 4-pentyn-2-ol
CC(CC#C)O
84.12
118.16
Odorless
25g
C5H8O
1756620
JTHLRRZARWSHBE-UHFFFAOYNA-N
pent-4-yn-2-ol
92915
≥98%

Chemical Identifiers

2117-11-5
84.12
JTHLRRZARWSHBE-UHFFFAOYNA-N
92915
CC(CC#C)O
C5H8O
MFCD00014472
4-pentyn-2-ol, 4-pentyne-2-ol, +--4-pentyn-2-ol, r,s-4-pentyn-2-ol, 4-hydroxypentyne, pentyne-4-ol, 4-pentin-2-ol, 1-methyl-3-butyn-1-ol, +/--4-pentyn-2-ol, 4-pentyn-2-ol
pent-4-yn-2-ol
Safety and Handling

Safety and Handling

GHS H Statement
H361-H302
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.

GHS P Statement
P280h-P330-P501a
Wear protective gloves/clothing.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : YR8250000

Documents