Please login to your online account to display your discounted pricing

Alfa Aesar™ N(alpha)-Fmoc-L-tryptophan, 98%

$21.55 - $57.86

Chemical Identifiers

Chemical Name or Material N(alpha)-Fmoc-L-tryptophan
CAS 35737-15-6
Molecular Formula C26H22N2O4
Formula Weight 426.48
MDL Number MFCD00037126
Beilstein 4216624
Synonym N(alpha)-9-Fluorenylmethoxycarbonyl-L-tryptophan; Fmoc-Trp-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2113003

  SDS
alfa aesar™
B2113003
1g Each for $21.55

AAB2113006

  SDS
alfa aesar™
B2113006
5g Each for $57.86
Description & Specifications

Specifications

Formula Weight 426.48
Chemical Name or Material N(alpha)-Fmoc-L-tryptophan
Assay 98%
Molecular Formula C26H22N2O4
MDL Number MFCD00037126
CAS 35737-15-6
Beilstein 4216624
Synonym N(alpha)-9-Fluorenylmethoxycarbonyl-L-tryptophan; Fmoc-Trp-OH
Melting Point 182° to 185°C
Quantity 1g
Optical Rotation −27° (c=1 in DMF)
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C