Please login to your online account to display your discounted pricing

Alfa Aesar™ N(alpha)-Fmoc-L-asparagine, 98%

$89.04 - $319.43

Chemical Identifiers

Chemical Name or Material N(alpha)-Fmoc-L-asparagine
CAS 71989-16-7
Molecular Formula C19H18N2O5
Formula Weight 354.36
MDL Number MFCD00037132
Beilstein 4335103
Synonym N(alpha)-9-Fluorenylmethoxycarbonyl-L-asparagine; Fmoc-Asn-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2100806

  SDS
alfa aesar™
B2100806
5g Each for $89.04

AAB2100814

  SDS
alfa aesar™
B2100814
25g Each for $319.43
Description & Specifications

Specifications

Formula Weight 354.36
Recommended Storage 2° to 8°C
Assay 98%
Beilstein 4335103
Optical Rotation −12±1° (c=1 in DMF)
Chemical Name or Material N(alpha)-Fmoc-L-asparagine
Quantity 5g
Molecular Formula C19H18N2O5
MDL Number MFCD00037132
Melting Point ≈188°C (dec.)
Synonym N(alpha)-9-Fluorenylmethoxycarbonyl-L-asparagine; Fmoc-Asn-OH
CAS 71989-16-7
Safety and Handling

2° to 8°C