Please login to your online account to display your discounted pricing

Alfa Aesar™ N(alpha)-Fmoc-D-tryptophan, 98%

$210.95 - $210.95

Chemical Identifiers

Chemical Name or Material N(alpha)-Fmoc-D-tryptophan
CAS 86123-11-7
Molecular Formula C26H22N2O4
Formula Weight 426.48
MDL Number MFCD00062954
Synonym N-(9-Fluorenylmethoxycarbonyl)-D-tryptophan; Fmoc-D-Trp-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2202206

  SDS
alfa aesar™
B2202206
5g Each for $210.95

AAB2202203

  SDS
alfa aesar™
B2202203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2202214

  SDS
alfa aesar™
B2202214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material N(alpha)-Fmoc-D-tryptophan
CAS 86123-11-7
Recommended Storage 2° to 8°C
Assay 98%
Molecular Formula C26H22N2O4
Formula Weight 426.48
MDL Number MFCD00062954
Synonym N-(9-Fluorenylmethoxycarbonyl)-D-tryptophan; Fmoc-D-Trp-OH
Melting Point 182° to 185°C
Quantity 5g
Safety and Handling

2° to 8°C