Please login to your online account to display your discounted pricing

Alfa Aesar™ N(alpha)-Ethoxycarbonyl-L-asparagine, 97%

$174.78 - $174.78

Chemical Identifiers

Chemical Name or Material N(alpha)-Ethoxycarbonyl-L-asparagine
CAS 16639-91-1
Molecular Formula C7H12N2O5
Formula Weight 204.18
MDL Number MFCD00082761
Beilstein 1712104
Synonym N(alpha)-Carbethoxy-L-asparagine; N(alpha)-Carboethoxy-L-asparagine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0932706

  SDS
alfa aesar™
L0932706
5g Each for $174.78
Description & Specifications

Specifications

Formula Weight 204.18
Chemical Name or Material N(alpha)-Ethoxycarbonyl-L-asparagine
MDL Number MFCD00082761
CAS 16639-91-1
Assay 97%
Synonym N(alpha)-Carbethoxy-L-asparagine; N(alpha)-Carboethoxy-L-asparagine
Recommended Storage 2° to 8°C
Quantity 5g
Beilstein 1712104
Melting Point ≈166°C
Molecular Formula C7H12N2O5
Safety and Handling

2° to 8°C