Please login to your online account to display your discounted pricing

Alfa Aesar™ N,alpha-Diphenyl nitrone, 97%

$48.64 - $526.43

Chemical Identifiers

Chemical Name or Material N,alpha-Diphenyl nitrone
CAS 1137-96-8
Molecular Formula C13H11NO
Formula Weight 197.24
MDL Number MFCD00013895
Beilstein 608470
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0018503

  SDS
alfa aesar™
L0018503
1g Each for $48.64

AAL0018506

  SDS
alfa aesar™
L0018506
5g Each for $140.39

AAL0018514

  SDS
alfa aesar™
L0018514
25g Each for $526.43
Description & Specifications

Specifications

Formula Weight 197.24
Chemical Name or Material N,alpha-Diphenyl nitrone
Assay 97%
Molecular Formula C13H11NO
MDL Number MFCD00013895
CAS 1137-96-8
Beilstein 608470
Recommended Storage 2° to 8°C
Melting Point 113° to 114°C
Quantity 1g

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in water at 25°C, 1120 mg/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

2° to 8°C