missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-α-BOC-N(im)-tosyl-L-histidine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC440860050

 View more versions of this product

Catalog No. AC440860050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

N-α-BOC-N(im)-tosyl-L-histidine
117.0°C to 118.0°C
35899-43-5
C18H23N3O6S
DCLJSEPKYJSEHW-HNNXBMFYSA-N
(2S)-3-[1-(4-methylphenyl)sulfonylimidazol-4-yl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
6455010
98%
+ 17.30
5g
97.5% min. (HPLC)
2s-2-tert-butoxycarbonyl amino-3-1-4-methylbenzenesulfonyl imidazol-4-yl propanoic acid, ambotzbaa1153, boc-his tos, boc-his tos-oh, boc-his-tos, boc-his-tos-oh, boc-l-his tos-oh, boc-l-histidine tosyl, pubchem7363, s-2-tert-butoxycarbonyl amino-3-1-tosyl-1h-imidazol-4-yl propanoic acid
CC1=CC=C(C=C1)S(=O)(=O)N2C=C(N=C2)CC(C(=O)O)NC(=O)OC(C)(C)C
409.46
409.46

Chemical Identifiers

35899-43-5
409.46
2s-2-tert-butoxycarbonyl amino-3-1-4-methylbenzenesulfonyl imidazol-4-yl propanoic acid, ambotzbaa1153, boc-his tos, boc-his tos-oh, boc-his-tos, boc-his-tos-oh, boc-l-his tos-oh, boc-l-histidine tosyl, pubchem7363, s-2-tert-butoxycarbonyl amino-3-1-tosyl-1h-imidazol-4-yl propanoic acid
(2S)-3-[1-(4-methylphenyl)sulfonylimidazol-4-yl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
C18H23N3O6S
DCLJSEPKYJSEHW-HNNXBMFYSA-N
6455010
CC1=CC=C(C=C1)S(=O)(=O)N2C=C(N=C2)CC(C(=O)O)NC(=O)OC(C)(C)C
SDS Documents