Please login to your online account to display your discounted pricing

N-alpha-BOC-N(im)-tosyl-L-histidine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-α-BOC-N(im)-tosyl-L-histidine
Assay Percent Range 98%
Molecular Formula C18H23N3O6S
Formula Weight 409.46
Synonym BOC-His(Tos)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC440860050

 
thermo fisher scientific
AC440860050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC440860250

 
thermo fisher scientific
AC440860250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 409.46
Quantity 5g
Chemical Name or Material N-α-BOC-N(im)-tosyl-L-histidine
Assay Percent Range 98%
Molecular Formula C18H23N3O6S
CAS 35899-43-5
Synonym BOC-His(Tos)-OH