Please login to your online account to display your discounted pricing

Alfa Aesar™ N(alpha)-Boc-L-tryptophan, 98+%

$85.58 - $317.93

Chemical Identifiers

Chemical Name or Material N(alpha)-Boc-L-tryptophan
CAS 13139-14-5
Molecular Formula C16H20N2O4
Formula Weight 304.35
MDL Number MFCD00065595
Beilstein 39677
Synonym Boc-Trp-OH; N(alpha)-tert-Butoxycarbonyl-L-tryptophan
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1602306

  SDS
alfa aesar™
A1602306
5g Each for $85.58

AAA1602314

  SDS
alfa aesar™
A1602314
25g Each for $317.93
Description & Specifications

Specifications

Formula Weight 304.35
Assay ≥98%
CAS 13139-14-5
Quantity 5g
Molecular Formula C16H20N2O4
Melting Point ≈138°C (dec.)
Recommended Storage 2° to 8°C
TSCA TSCA
MDL Number MFCD00065595
Chemical Name or Material N(alpha)-Boc-L-tryptophan
Beilstein 39677
Synonym Boc-Trp-OH; N(alpha)-tert-Butoxycarbonyl-L-tryptophan
Optical Rotation −19.5° (c=1 in DMF)
Safety and Handling

2° to 8°C