Please login to your online account to display your discounted pricing

Alfa Aesar™ N(alpha)-Boc-L-histidine, 98+%

$33.91 - $33.91

Chemical Identifiers

Chemical Name or Material N(alpha)-Boc-L-histidine
CAS 17791-52-5
Molecular Formula C11H17N3O4
Formula Weight 255.27
MDL Number MFCD00065576
Beilstein 755289
Synonym Boc-His-OH; N(alpha)-tert-Butoxycarbonyl-L-histidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2204203

  SDS
alfa aesar™
B2204203
1g Each for $33.91

AAB2204206

  SDS
alfa aesar™
B2204206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material N(alpha)-Boc-L-histidine
CAS 17791-52-5
Assay ≥98%
Molecular Formula C11H17N3O4
Formula Weight 255.27
MDL Number MFCD00065576
Beilstein 755289
Synonym Boc-His-OH; N(alpha)-tert-Butoxycarbonyl-L-histidine
Melting Point ≈198°C (dec.)
Quantity 1g
Optical Rotation +26° (c=1 in Methanol)