Please login to your online account to display your discounted pricing

Alfa Aesar™ N(alpha)-Boc-L-glutamine, 98+%

$71.57 - $212.46

Chemical Identifiers

Chemical Name or Material N(alpha)-Boc-L-glutamine
CAS 13726-85-7
Molecular Formula C10H18N2O5
Formula Weight 246.26
MDL Number MFCD00065571
Beilstein 2127805
Synonym Boc-Gln-OH; N(alpha)-tert-Butoxycarbonyl-L-glutamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0860406

  SDS
alfa aesar™
L0860406
5g Each for $71.57

AAL0860414

  SDS
alfa aesar™
L0860414
25g Each for $212.46
Description & Specifications

Specifications

Formula Weight 246.26
Chemical Name or Material N(alpha)-Boc-L-glutamine
Synonym Boc-Gln-OH; N(alpha)-tert-Butoxycarbonyl-L-glutamine
Molecular Formula C10H18N2O5
Assay ≥98%
Recommended Storage 2° to 8°C
MDL Number MFCD00065571
Optical Rotation −18.5° (c=1, DMF)
Beilstein 2127805
TSCA TSCA
Melting Point 118° to 120°C
CAS 13726-85-7
Quantity 5g

Applications
It is employed in the synthesis and analgesic activity of human .beta.-endorphin.

Solubility
Soluble in DMSO and methanol. Soluble in 1 mmole in 2 ml DMF.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

2° to 8°C