missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N(alpha)-Boc-L-arginine, 98%

Manufacturer:  Alfa Aesar™ H5337903

 View more versions of this product

Catalog No. AAH5337903



Specifications

Specifications

N(alpha)-Boc-L-arginine
−6.5° (c=1 in Acetic Acid)
13726-76-6
MFCD00042632
boc-arg-oh, boc-arginine, s-2-tert-butoxycarbonyl amino-5-guanidinopentanoic acid, boc-l-arg-oh, n2-tert-butoxycarbonyl-l-arginine, n-boc-l-arginine, na-tert-butoxycarbonyl-l-arginine, n-t-boc-l-arginine, n alpha-boc-l-arginine, n~2~-tert-butoxycarbonyl-l-arginine
CC(C)(C)OC(=O)NC(CCCN=C(N)N)C(=O)O
274.321
274.32
113°C (235°F)
1g
C11H22N4O4
2419628
HSQIYOPBCOPMSS-ZETCQYMHSA-N
(2S)-5-(diaminomethylideneamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
114667
98%

Chemical Identifiers

13726-76-6
274.321
HSQIYOPBCOPMSS-ZETCQYMHSA-N
114667
CC(C)(C)OC(=O)NC(CCCN=C(N)N)C(=O)O
C11H22N4O4
MFCD00042632
boc-arg-oh, boc-arginine, s-2-tert-butoxycarbonyl amino-5-guanidinopentanoic acid, boc-l-arg-oh, n2-tert-butoxycarbonyl-l-arginine, n-boc-l-arginine, na-tert-butoxycarbonyl-l-arginine, n-t-boc-l-arginine, n alpha-boc-l-arginine, n~2~-tert-butoxycarbonyl-l-arginine
(2S)-5-(diaminomethylideneamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SDS Documents