Please login to your online account to display your discounted pricing

Alfa Aesar™ N(alpha)-Boc-L-2,3-diaminopropionic acid, 97%

$88.15 - $88.15

Chemical Identifiers

Chemical Name or Material N(alpha)-Boc-L-2,3-diaminopropionic acid
CAS 73259-81-1
Molecular Formula C8H16N2O4
Formula Weight 204.23
MDL Number MFCD00236843
Synonym Boc-Dap-OH; 3-Amino-N-Boc-L-alanine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2691903

  SDS
alfa aesar™
H2691903
1g Each for $88.15
Description & Specifications

Specifications

Formula Weight 204.23
CAS 73259-81-1
Optical Rotation −14° (c=1 in Water)
Melting Point ≈210°C
Chemical Name or Material N(alpha)-Boc-L-2,3-diaminopropionic acid
Assay 97%
Quantity 1g
Molecular Formula C8H16N2O4
MDL Number MFCD00236843
Synonym Boc-Dap-OH; 3-Amino-N-Boc-L-alanine