Please login to your online account to display your discounted pricing

Alfa Aesar™ N(alpha)-Boc-D-glutamine, 98+%

$27.88 - $71.12

Chemical Identifiers

Chemical Name or Material N(alpha)-Boc-D-glutamine
CAS 61348-28-5
Molecular Formula C10H18N2O5
Formula Weight 246.26
MDL Number MFCD00038158
Beilstein 1981311
Synonym Boc-D-Gln-OH; N(alpha)-tert-Butoxycarbonyl-D-glutamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0853603

  SDS
alfa aesar™
L0853603
1g Each for $71.12

AAL08536MD

  SDS
alfa aesar™
L08536MD
250mg Each for $27.88
Description & Specifications

Specifications

Formula Weight 246.26
Chemical Name or Material N(alpha)-Boc-D-glutamine
Assay ≥98%
Molecular Formula C10H18N2O5
MDL Number MFCD00038158
CAS 61348-28-5
Synonym Boc-D-Gln-OH; N(alpha)-tert-Butoxycarbonyl-D-glutamine
Beilstein 1981311
Melting Point 117° to 119°C
Quantity 1g
Optical Rotation +3.5° (c=2 in Ethanol)
Recommended Storage 2° to 8°C

Applications
N2-Boc-D-glutamine is an N-Boc-protected form of D-Glutamine. D-Glutamine is an unnatural isomer of L-Glutamine that is present in human plasma an is a source of liberated ammonia. D-Glutamine can be synthesized by enzymatic means or can be found in cheeses, wine and vinegars as well.

Solubility
Soluble, 1mmol in 2mL DMF. Soluble in chloroform and methanol.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.

Safety and Handling

2° to 8°C