Please login to your online account to display your discounted pricing

Alfa Aesar™ N(alpha)-Benzyloxycarbonyl-L-tryptophan, 98+%

$32.95 - $99.14

Chemical Identifiers

Chemical Name or Material N(alpha)-Benzyloxycarbonyl-L-tryptophan
CAS 7432-21-5
Molecular Formula C19H18N2O4
Formula Weight 338.37
MDL Number MFCD00065700
Beilstein 96743
Synonym N(alpha)-Cbz-L-tryptophan; Z-Trp-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0753506

 
alfa aesar™
L0753506
5g Each for $32.95

AAL0753514

 
alfa aesar™
L0753514
25g Each for $99.14
Description & Specifications

Specifications

Formula Weight 338.37
Chemical Name or Material N(alpha)-Benzyloxycarbonyl-L-tryptophan
Assay ≥98%
Molecular Formula C19H18N2O4
CAS 7432-21-5
Synonym N(alpha)-Cbz-L-tryptophan; Z-Trp-OH
MDL Number MFCD00065700
Beilstein 96743
Melting Point 121° to 126°C
Quantity 5g
Odor Odorless
Optical Rotation +3.1° (c=3 in Acetic Acid)
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C