Please login to your online account to display your discounted pricing

Alfa Aesar™ N(alpha)-Benzyloxycarbonyl-L-asparagine, 98+%

$34.20 - $104.42

Chemical Identifiers

Chemical Name or Material N(alpha)-Benzyloxycarbonyl-L-asparagine
CAS 2304-96-3
Molecular Formula C12H14N2O5
Formula Weight 266.26
MDL Number MFCD00008035
Beilstein 3085452
Synonym N(alpha)-Cbz-L-asparagine; Z-Asn-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0859206

  SDS
alfa aesar™
L0859206
5g Each for $34.20

AAL0859214

  SDS
alfa aesar™
L0859214
25g Each for $104.42
Description & Specifications

Specifications

Formula Weight 266.26
MDL Number MFCD00008035
Chemical Name or Material N(alpha)-Benzyloxycarbonyl-L-asparagine
Beilstein 3085452
Synonym N(alpha)-Cbz-L-asparagine; Z-Asn-OH
Assay ≥98%
CAS 2304-96-3
TSCA TSCA
Quantity 5g
Melting Point 162° to 166°C
Optical Rotation +6° (c=2 in Acetic Acid)
Molecular Formula C12H14N2O5