Please login to your online account to display your discounted pricing

Alfa Aesar™ N(alpha)-Benzyloxycarbonyl-L-arginine, 99%

$34.20 - $442.99

Chemical Identifiers

Chemical Name or Material N(alpha)-Benzyloxycarbonyl-L-arginine
CAS 1234-35-1
Molecular Formula C14H20N4O4
Formula Weight 308.34
MDL Number MFCD00001762
Beilstein 2169267
Synonym N(alpha)-Cbz-L-arginine; Z-Arg-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1704306

  SDS
alfa aesar™
A1704306
5g Each for $34.20

AAA1704314

  SDS
alfa aesar™
A1704314
25g Each for $140.58

AAA1704322

  SDS
alfa aesar™
A1704322
100g Each for $442.99
Description & Specifications

Specifications

Formula Weight 308.34
Beilstein 2169267
Optical Rotation −9.5° (c=5 in 0.2N HCl)
TSCA TSCA
Melting Point 173° to 175°C
Quantity 5g
MDL Number MFCD00001762
CAS 1234-35-1
Synonym N(alpha)-Cbz-L-arginine; Z-Arg-OH
Molecular Formula C14H20N4O4
Chemical Name or Material N(alpha)-Benzyloxycarbonyl-L-arginine
Assay 99%