Please login to your online account to display your discounted pricing

Alfa Aesar™ N(alpha)-Benzyloxycarbonyl-D-tryptophan, 98+%

$33.59 - $88.59

Chemical Identifiers

Chemical Name or Material N(alpha)-Benzyloxycarbonyl-D-tryptophan
CAS 2279-15-4
Molecular Formula C19H18N2O4
Formula Weight 338.37
MDL Number MFCD00037945
Beilstein 96744
Synonym N(alpha)-Cbz-D-tryptophan; Z-D-Trp-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL08769MD

  SDS
alfa aesar™
L08769MD
250mg Each for $33.59

AAL0876903

  SDS
alfa aesar™
L0876903
1g Each for $88.59
Description & Specifications

Specifications

Chemical Name or Material N(alpha)-Benzyloxycarbonyl-D-tryptophan
CAS 2279-15-4
Recommended Storage 2° to 8°C
Assay ≥98%
Molecular Formula C19H18N2O4
Formula Weight 338.37
MDL Number MFCD00037945
Beilstein 96744
Synonym N(alpha)-Cbz-D-tryptophan; Z-D-Trp-OH
Melting Point 122° to 124°C
Quantity 250mg
Optical Rotation −3.1° (c=3 in Acetic Acid)
Safety and Handling

2° to 8°C