Please login to your online account to display your discounted pricing

Alfa Aesar™ N(alpha)-Benzyloxycarbonyl-D-histidine, 98+%

$112.51 - $112.51

Chemical Identifiers

Chemical Name or Material N(alpha)-Benzyloxycarbonyl-D-histidine
CAS 67424-93-5
Molecular Formula C14H15N3O4
Formula Weight 289.29
MDL Number MFCD00067702
Beilstein 92342
Synonym N(alpha)-Cbz-D-histidine; Z-D-His-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL09005MD

 
alfa aesar™
L09005MD
250mg Each for $112.51
Description & Specifications

Specifications

Formula Weight 289.29
TSCA TSCA
Assay ≥98%
Molecular Formula C14H15N3O4
MDL Number MFCD00067702
CAS 67424-93-5
Synonym N(alpha)-Cbz-D-histidine; Z-D-His-OH
Quantity 250mg
Recommended Storage 2° to 8°C
Chemical Name or Material N(alpha)-Benzyloxycarbonyl-D-histidine
Beilstein 92342
Melting Point 161° to 163°C
Optical Rotation +24° (c=6 in 6N HCl)
Safety and Handling

2° to 8°C