missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-[alpha-(4-Chlorophenyl)benzyl]formamide, 98%, Alfa Aesar™

$70.39 - $208.93

Chemical Identifiers

CAS 202414-68-4
Molecular Formula C14H12ClNO
Molecular Weight (g/mol) 245.706
InChI Key HDGOSEHHSVUCBO-UHFFFAOYSA-N
Synonym formamide, n-4-chlorophenyl phenylmethyl
PubChem CID 10377235
IUPAC Name N-[(4-chlorophenyl)-phenylmethyl]formamide
SMILES C1=CC=C(C=C1)C(C2=CC=C(C=C2)Cl)NC=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH61366MD View Documents Thermo Fisher Scientific
H61366MD
250mg Each for $70.39
 
AAH6136603 View Documents Thermo Fisher Scientific
H6136603
1g Each for $208.93
 
Specifications

Chemical Identifiers

202414-68-4
245.706
formamide, n-4-chlorophenyl phenylmethyl
N-[(4-chlorophenyl)-phenylmethyl]formamide
C14H12ClNO
HDGOSEHHSVUCBO-UHFFFAOYSA-N
10377235
C1=CC=C(C=C1)C(C2=CC=C(C=C2)Cl)NC=O

Specifications

250mg
C14H12ClNO
HDGOSEHHSVUCBO-UHFFFAOYSA-N
N-[(4-chlorophenyl)-phenylmethyl]formamide
10377235
Solid
N-[alpha-(4-Chlorophenyl)benzyl]formamide
202414-68-4
formamide, n-4-chlorophenyl phenylmethyl
C1=CC=C(C=C1)C(C2=CC=C(C=C2)Cl)NC=O
245.706
245.7
98%
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures