missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-[alpha-(4-Chlorophenyl)benzyl]formamide, 98%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H6136603

 View more versions of this product

Catalog No. AAH6136603



Specifications

Specifications

N-[alpha-(4-Chlorophenyl)benzyl]formamide
202414-68-4
formamide, n-4-chlorophenyl phenylmethyl
C1=CC=C(C=C1)C(C2=CC=C(C=C2)Cl)NC=O
245.706
245.7
98%
1g
C14H12ClNO
HDGOSEHHSVUCBO-UHFFFAOYSA-N
N-[(4-chlorophenyl)-phenylmethyl]formamide
10377235
Solid

Chemical Identifiers

202414-68-4
245.706
formamide, n-4-chlorophenyl phenylmethyl
N-[(4-chlorophenyl)-phenylmethyl]formamide
C14H12ClNO
HDGOSEHHSVUCBO-UHFFFAOYSA-N
10377235
C1=CC=C(C=C1)C(C2=CC=C(C=C2)Cl)NC=O
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents