missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acryloylsarcosine methyl ester resin, ca 1 mmol/g

$435.62 - $435.62

Chemical Identifiers

MDL Number MFCD03792464
Synonym Sheppard acrylamide resin
Chemical Name or Material N-Acryloylsarcosine methyl ester resin
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1959303 SDS Alfa Aesar™
L1959303
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAL1959306 SDS Alfa Aesar™
L1959306
5g Each for $435.62
Specifications Description & Specifications

Chemical Identifiers

MFCD03792464
Sheppard acrylamide resin
N-Acryloylsarcosine methyl ester resin

Specifications

≈1mmol/g
MFCD03792464
N-Acryloylsarcosine methyl ester resin
1g
Sheppard acrylamide resin