Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acryloylsarcosine methyl ester, 97%, stab. with 250ppm 4-methoxyphenol

$106.99 - $397.78

Chemical Identifiers

Chemical Name or Material N-Acryloylsarcosine methyl ester
CAS 72065-23-7
Name Note stabilized with 250ppm 4-methoxyphenol
Molecular Formula C7H11NO3
Formula Weight 157.17
MDL Number MFCD00151792
Beilstein 4964477
Synonym Methyl N-methyl-N-(1-oxo-2-propenyl)glycinate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1312804

  SDS
alfa aesar™
L1312804
2g Each for $106.99

AAL1312809

  SDS
alfa aesar™
L1312809
10g Each for $397.78
Description & Specifications

Specifications

Formula Weight 157.17
Chemical Name or Material N-Acryloylsarcosine methyl ester
Health Hazard 1 Warning
Assay 97%
CAS 72065-23-7
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Synonym Methyl N-methyl-N-(1-oxo-2-propenyl)glycinate
Quantity 2g
Name Note stabilized with 250ppm 4-methoxyphenol
Flash Point >110°C (230°F)
MDL Number MFCD00151792
Boiling Point 96°C (0.4mmHg)
Beilstein 4964477
Molecular Formula C7H11NO3
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Recommended Storage 2° to 8°C
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

2° to 8°C