N-Acetyltryptamine, Tocris
Melatonin partial agonist (MT1/MT2); Also MT3 antagonist
Manufacturer:  R & D Systems 035710
 View more versions of this product
Catalog No. 035710
Specifications Description & Specifications
Specifications
N-Acetyltryptamine
>99%
202.26
1016-47-3
C12H14N2O
10mg
Chemical Identifiers
>99%
C12H14N2O
202.26
N-Acetyltryptamine
Safety and Handling
Recommended Storage : Store at Room Temperature