Please login to your online account to display your discounted pricing

N-Acetyltryptamine, Tocris

Melatonin partial agonist (MT1/MT2); Also MT3 antagonist

$146.74 - $618.87

Chemical Identifiers

Chemical Name or Material N-Acetyltryptamine
Assay Percent Range >99%
Molecular Formula C12H14N2O
Formula Weight 202.26
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

035710

 
r & d systems
035710
10mg Each for $146.74

035750

 
r & d systems
035750
50mg Each for $618.87
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 202.26
Assay Percent Range >99%
Chemical Name or Material N-Acetyltryptamine
CAS 1016-47-3
Recommended Storage Store at Room Temperature
Molecular Formula C12H14N2O
Safety and Handling

Store at Room Temperature