Please login to your online account to display your discounted pricing

N-Acetyltryptamine, Tocris


Melatonin partial agonist (MT1/MT2); Also MT3 antagonist

Quantity 50mg

Manufacturer: r & d systems  035750

 View more versions of this product

Catalog No. 035750

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 202.26
CAS 1016-47-3
Molecular Formula C12H14N2O
Recommended Storage Store at Room Temperature
Chemical Name or Material N-Acetyltryptamine
Assay Percent Range >99%

Specifications

Chemical Name or Material N-Acetyltryptamine
Assay Percent Range >99%
Molecular Formula C12H14N2O
Formula Weight 202.26
Accessories

Store at Room Temperature