Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetylthiourea, 99+%

Chemical Identifiers

Chemical Name or Material N-Acetylthiourea
CAS 591-08-2
Molecular Formula C3H6N2OS
Formula Weight 118.16
MDL Number MFCD00004937
UN Number UN2811
Beilstein 969960
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2119814

 
alfa aesar™
B2119814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2119822

 
alfa aesar™
B2119822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2119836

 
alfa aesar™
B2119836
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 118.16
Chemical Name or Material N-Acetylthiourea
TSCA TSCA
Assay ≥99%
Molecular Formula C3H6N2OS
MDL Number MFCD00004937
CAS 591-08-2
UN Number UN2811
Beilstein 969960
Health Hazard 2 GHS H Statement
H300
Fatal if swallowed.
Health Hazard 3 GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number YR7700000
Odor Odorless
Melting Point 165° to 169°C
Quantity 25g
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H300
Fatal if swallowed.