missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetylthiourea, 98%

Manufacturer:  Alfa Aesar™ 4700822

 View more versions of this product

Catalog No. AA4700822



Specifications

Specifications

N-Acetylthiourea
Cream
100g
C3H6N2OS
UN2811
n-acetylthiourea, acetylthiourea, 1-acetylthiourea, 1-acetyl-2-thiourea, acetothiourea, n-acetyl-2-thiourea, acetamide, n-aminothioxomethyl, n-acetylthiocarbamide, urea, 1-acetyl-2-thio, acetylthiocarbamide
CC(=O)NC(=S)N
118.154
118.16
98%
Odorless
165°C to 169°C
591-08-2
MFCD00004937
969960
IPCRBOOJBPETMF-UHFFFAOYSA-N
N-carbamothioylacetamide
2723593
Crystalline Powder

Chemical Identifiers

591-08-2
118.154
IPCRBOOJBPETMF-UHFFFAOYSA-N
2723593
CC(=O)NC(=S)N
C3H6N2OS
MFCD00004937
n-acetylthiourea, acetylthiourea, 1-acetylthiourea, 1-acetyl-2-thiourea, acetothiourea, n-acetyl-2-thiourea, acetamide, n-aminothioxomethyl, n-acetylthiocarbamide, urea, 1-acetyl-2-thio, acetylthiocarbamide
N-carbamothioylacetamide
Safety and Handling

Safety and Handling

GHS H Statement
H300
Fatal if swallowed.

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : YR7700000

TSCA : TSCA

SDS Documents