Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetylphthalimide, 97%

$62.49 - $260.14

Chemical Identifiers

Chemical Name or Material N-Acetylphthalimide
CAS 1971-49-9
Molecular Formula C10H7NO3
Formula Weight 189.17
MDL Number MFCD00023054
Beilstein 144637
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1485506

  SDS
alfa aesar™
A1485506
5g Each for $62.49

AAA1485514

  SDS
alfa aesar™
A1485514
25g Each for $260.14
Description & Specifications

Specifications

Formula Weight 189.17
Chemical Name or Material N-Acetylphthalimide
Assay 97%
Molecular Formula C10H7NO3
MDL Number MFCD00023054
CAS 1971-49-9
Beilstein 144637
Melting Point 135° to 139°C
Quantity 5g