missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetylphthalimide, 97%

Manufacturer:  Alfa Aesar™ A1485514

 View more versions of this product

Catalog No. AAA1485514



Specifications

Specifications

N-Acetylphthalimide
25g
C10H7NO3
144637
INZUQGFQRYAKQQ-UHFFFAOYSA-N
2-acetylisoindole-1,3-dione
243267
97%
135°C to 139°C
1971-49-9
MFCD00023054
n-acetylphthalimide, 2-acetylisoindoline-1,3-dione, acetylphthalimide, phthalimide, n-acetyl, 2-acetyl-1,3-dioxobenzo c azoline, n-acetylphthali-mide, 1h-isoindole-1,3 2h-dione, 2-acetyl, maybridge1_007322, acmc-209f0t, n-acetylphthalimide
CC(=O)N1C(=O)C2=CC=CC=C2C1=O
189.17
189.17

Chemical Identifiers

1971-49-9
189.17
INZUQGFQRYAKQQ-UHFFFAOYSA-N
243267
CC(=O)N1C(=O)C2=CC=CC=C2C1=O
C10H7NO3
MFCD00023054
n-acetylphthalimide, 2-acetylisoindoline-1,3-dione, acetylphthalimide, phthalimide, n-acetyl, 2-acetyl-1,3-dioxobenzo c azoline, n-acetylphthali-mide, 1h-isoindole-1,3 2h-dione, 2-acetyl, maybridge1_007322, acmc-209f0t, n-acetylphthalimide
2-acetylisoindole-1,3-dione
SDS Documents