missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-(-)-Acetylneuraminic acid, 97%

Manufacturer:  Alfa Aesar™ L13950MC

 View more versions of this product

Catalog No. AAL13950MC



Description

Description

Applications
N-Acetylneuraminic acid is useful biologically in neurotransmission, leukocyte extravasation, viral or bacterial infections and carbohydrate-protein recognition. Neu5Ac is used to study its biochemistry, metabolism and uptake in vivo and in vitro. Neu5Ac is used in the development of nanocarriers.

Solubility
Soluble in water (50 mg/ml).

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.
Specifications

Specifications

N-(-)-Acetylneuraminic acid
~185°C
131-48-6
MFCD00006620
14,8484
SQVRNKJHWKZAKO-LFIUDZTESA-N
(4S,5S,6R)-5-acetamido-2,4-dihydroxy-6-[(1R,2R)-1,2,3-trihydroxypropyl]oxane-2-carboxylic acid
126963458
97%
−32° (c=0.5 in Water)
100mg
C11H19NO9
1716283
n-acetylneuramic acid, spectrum_000450, spectrum2_000559, spectrum4_000432, spectrum5_001326, spectrum2300147, 2s,4s,5s,6s-5-acetamido-2,4-dihydroxy-6-2r-1,2,3-trihydroxypropyl oxane-2-carboxylic acid
CC(=O)NC1C(CC(OC1C(C(CO)O)O)(C(=O)O)O)O
309.271
309.28

Chemical Identifiers

131-48-6
309.271
SQVRNKJHWKZAKO-LFIUDZTESA-N
126963458
CC(=O)NC1C(CC(OC1C(C(CO)O)O)(C(=O)O)O)O
C11H19NO9
MFCD00006620
n-acetylneuramic acid, spectrum_000450, spectrum2_000559, spectrum4_000432, spectrum5_001326, spectrum2300147, 2s,4s,5s,6s-5-acetamido-2,4-dihydroxy-6-2r-1,2,3-trihydroxypropyl oxane-2-carboxylic acid
(4S,5S,6R)-5-acetamido-2,4-dihydroxy-6-[(1R,2R)-1,2,3-trihydroxypropyl]oxane-2-carboxylic acid
Safety and Handling

Safety and Handling

TSCA : TSCA

Recommended Storage : −30° to −10°C

Documents