Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetylmorpholine, 98%

$124.16 - $124.16

Chemical Identifiers

Chemical Name or Material N-Acetylmorpholine
CAS 1696-20-4
Molecular Formula C6H11NO2
Formula Weight 129.16
MDL Number MFCD00006171
Beilstein 111971
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2412022

  SDS
alfa aesar™
B2412022
100g Each for $124.16
Description & Specifications

Specifications

Formula Weight 129.16
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 1696-20-4
Quantity 100g
Boiling Point 245° to 246°C
Chemical Name or Material N-Acetylmorpholine
Beilstein 111971
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flash Point >110°C (230°F)
Melting Point 9° to 10°C
MDL Number MFCD00006171
Molecular Formula C6H11NO2
RTECS Number QD7000000
TSCA TSCA
Assay 98%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.