missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetylglycylglycine, 97%

$90.68 - $289.92

Chemical Identifiers

CAS 5687-48-9
Molecular Formula C6H10N2O4
Molecular Weight (g/mol) 174.156
MDL Number MFCD00037788
InChI Key ZCASHLUDUSAKNN-UHFFFAOYSA-N
Synonym 2-2-acetamidoacetamido acetic acid, n-acetylglycylglycine, ac-gly-gly-oh, unii-01u1cr2e6l, 2-acetamidoacetamido acetic acid, n-ac-gly-gly, dpgt, acetyl-gly-gly, acetyl-glycylglycine, glycine, n-n-acetylglycyl
PubChem CID 97591
IUPAC Name 2-[(2-acetamidoacetyl)amino]acetic acid
SMILES CC(=O)NCC(=O)NCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH65178MD View Documents Alfa Aesar™
H65178MD
250mg Each for $90.68
 
AAH6517803 View Documents Alfa Aesar™
H6517803
1g Each for $289.92
 
Specifications

Chemical Identifiers

5687-48-9
174.156
ZCASHLUDUSAKNN-UHFFFAOYSA-N
97591
CC(=O)NCC(=O)NCC(=O)O
C6H10N2O4
MFCD00037788
2-2-acetamidoacetamido acetic acid, n-acetylglycylglycine, ac-gly-gly-oh, unii-01u1cr2e6l, 2-acetamidoacetamido acetic acid, n-ac-gly-gly, dpgt, acetyl-gly-gly, acetyl-glycylglycine, glycine, n-n-acetylglycyl
2-[(2-acetamidoacetyl)amino]acetic acid

Specifications

5687-48-9
MFCD00037788
2-2-acetamidoacetamido acetic acid, n-acetylglycylglycine, ac-gly-gly-oh, unii-01u1cr2e6l, 2-acetamidoacetamido acetic acid, n-ac-gly-gly, dpgt, acetyl-gly-gly, acetyl-glycylglycine, glycine, n-n-acetylglycyl
CC(=O)NCC(=O)NCC(=O)O
174.156
174.16
97%
N-Acetylglycylglycine
C6H10N2O4
1782002
ZCASHLUDUSAKNN-UHFFFAOYSA-N
2-[(2-acetamidoacetyl)amino]acetic acid
97591
Solid
250mg