Please login to your online account to display your discounted pricing

N-Acetylglycyl-D-glutamic acid, Tocris

Potent convulsant

$21.11 - $21.11

Chemical Identifiers

Chemical Name or Material N-Acetylglycyl-D-glutamic acid
Assay Percent Range >98%
Molecular Formula C9H14N2O6
Formula Weight 246.22
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

03-951-0

 
jg finneran associates
554011G
10mg Pack of 100 Each for $21.11
Case of 10 Pack for $211.10
Description & Specifications

Specifications

Chemical Name or Material N-Acetylglycyl-D-glutamic acid
CAS 135701-69-8
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Molecular Formula C9H14N2O6
Formula Weight 246.22
Quantity 10mg
Safety and Handling

Store at Room Temperature