missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Acetylglycine 99%, ACROS Organics™

Manufacturer:  Acros Organics AC102570025

 View more versions of this product

Catalog No. AC102570025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N-Acetylglycine
0.5% max.
0.1% max.
207.0°C to 209.0°C
99%
98.5
99%
CH3CONHCH2CO2H
04, 354
ac-gly-oh
OKJIRPAQVSHGFK-UHFFFAOYSA-N
2-acetamidoacetic acid
10972
117.1
99%
Authentic
Plastic bottle
White
2.5kg
543-24-8
100.0
C4H7NO3
MFCD00004275
15, 75
Solubility in water: 2.7% (15°C). Other solubilities: moderately soluble in alcohol, slightly soluble in acetone, chloroform and, glacial acetic acid, practically insoluble in ether and benzene
CC(=O)NCC(=O)O
117.1
CHEBI:40410
Crystalline Powder

Chemical Identifiers

543-24-8
117.1
OKJIRPAQVSHGFK-UHFFFAOYSA-N
10972
2-acetamidoacetic acid
C4H7NO3
MFCD00004275
ac-gly-oh
CHEBI:40410
CC(=O)NCC(=O)O
Safety and Handling

Safety and Handling

EINECSNumber : 208-839-6

TSCA : TSCA

SDS Documents