Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetylglycinamide, 97%

$68.55 - $779.00

Chemical Identifiers

Chemical Name or Material N-Acetylglycinamide
CAS 2620-63-5
Molecular Formula CH3CONHCH2CONH2
Formula Weight 116.12
MDL Number MFCD00008029
Beilstein 1753982
Synonym 2-Acetamidoacetamide; Ac-Gly-NH_2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2494003

  SDS
alfa aesar™
B2494003
1g Each for $68.55

AAB2494006

  SDS
alfa aesar™
B2494006
5g Each for $226.01

AAB2494014

  SDS
alfa aesar™
B2494014
25g Each for $779.00
Description & Specifications

Specifications

Formula Weight 116.12
Chemical Name or Material N-Acetylglycinamide
Assay 97%
Molecular Formula CH3CONHCH2CONH2
MDL Number MFCD00008029
CAS 2620-63-5
Beilstein 1753982
Synonym 2-Acetamidoacetamide; Ac-Gly-NH_2
RTECS Number MB7507000
Odor Odorless
Melting Point 136° to 140°C
Quantity 1g