missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Acetylglycinamide, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2494006
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-Acetylglycinamide | |
136°C to 140°C | |
C4H8N2O2 | |
5 g | |
n-acetylglycine amide, n-alpha-acetylglycinamide, unii-97sv9afw2z, acetamide, n-2-amino-2-oxoethyl, nalpha-acetylglycinamide, n-.alpha.-acetylglycinamide, 97sv9afw2z, ethanimidic acid, n-2-amino-2-oxoethyl, acetylglutamide, acetylglycinamide | |
CC(=O)NCC(N)=O | |
116.12 | |
116.12 |
2620-63-5 | |
Odorless | |
MFCD00008029 | |
1753982 | |
WQELDIQOHGAHEM-UHFFFAOYSA-N | |
2-acetamidoacetamide | |
28326 | |
97% |
Chemical Identifiers
2620-63-5 | |
116.12 | |
WQELDIQOHGAHEM-UHFFFAOYSA-N | |
28326 | |
CC(=O)NCC(N)=O |
C4H8N2O2 | |
MFCD00008029 | |
n-acetylglycine amide, n-alpha-acetylglycinamide, unii-97sv9afw2z, acetamide, n-2-amino-2-oxoethyl, nalpha-acetylglycinamide, n-.alpha.-acetylglycinamide, 97sv9afw2z, ethanimidic acid, n-2-amino-2-oxoethyl, acetylglutamide, acetylglycinamide | |
2-acetamidoacetamide |
Safety and Handling
EINECSNumber : 220-058-2
RTECSNumber : MB7507000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only