missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Acetylethanolamine, ACROS Organics™

Manufacturer:  Acros Organics 146201000

 View more versions of this product

Catalog No. AC146201000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N-Acetylethanolamine
Glass bottle
White to Yellow
142-26-7
100.0
C4H9NO2
MFCD00002836
n-2-hydroxyethyl acetamide, n-acetylethanolamine, 2-acetamidoethanol, acetamide, n-2-hydroxyethyl, acetylcolamine, 2-acetylaminoethanol, acetamide mea, n-ethanolacetamide, n-acetyl-2-aminoethanol, hydroxyethyl acetamide
CC(=O)NCCO
103.121
CHEBI:74687
Liquid
Authentic
1.4720 to 1.4740 (20°C, 589nm)
100g
95.0
96%
CH3CONHCH2CH2OH
04, I, 430
PVCJKHHOXFKFRP-UHFFFAOYSA-N
N-(2-hydroxyethyl)acetamide
8880
103.12
≥95%

Chemical Identifiers

142-26-7
103.121
PVCJKHHOXFKFRP-UHFFFAOYSA-N
8880
N-(2-hydroxyethyl)acetamide
C4H9NO2
MFCD00002836
n-2-hydroxyethyl acetamide, n-acetylethanolamine, 2-acetamidoethanol, acetamide, n-2-hydroxyethyl, acetylcolamine, 2-acetylaminoethanol, acetamide mea, n-ethanolacetamide, n-acetyl-2-aminoethanol, hydroxyethyl acetamide
CHEBI:74687
CC(=O)NCCO
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 205-530-8

RTECSNumber : AC3120000

TSCA : TSCA

SDS Documents