Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetylethanolamine, 90+%

$52.88 - $137.42

Chemical Identifiers

Chemical Name or Material N-Acetylethanolamine
CAS 142-26-7
Molecular Formula C4H9NO2
Linear Formula CH3CONHCH2CH2OH
Formula Weight 103.12
MDL Number MFCD00002836
Synonym 2-Acetamidoethanol; N-(2-Hydroxyethyl)acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1863822

  SDS
alfa aesar™
A1863822
100g Each for $52.88

AAA1863836

  SDS
alfa aesar™
A1863836
500g Each for $137.42
Description & Specifications

Specifications

Formula Weight 103.12
Chemical Name or Material N-Acetylethanolamine
TSCA TSCA
Assay ≥90%
Molecular Formula C4H9NO2
Linear Formula CH3CONHCH2CH2OH
MDL Number MFCD00002836
CAS 142-26-7
Synonym 2-Acetamidoethanol; N-(2-Hydroxyethyl)acetamide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H318
Causes serious eye damage.
Boiling Point 151° to 155°C (5mmHg)
Quantity 100g
Flash Point 177°C (351°F)
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
RTECS Number AC3120000

Applications
It finds its uses as antistatic agent, foam boosting agent, hair conditioning humectant, skin conditioning surfactant and viscosity controlling agent in cosmetics industry. It is a non-tacky glycerine alternative for use in skin and hair care. It has humectant and lubricant properties for use in shampoos, treatment products and hair conditioners.

Solubility
Soluble in water, 1e+006 mg/L @ 25°C (est), soluble in alcohol and ether.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H318
Causes serious eye damage.