Please login to your online account to display your discounted pricing

N-Acetyl-Val-Asp-Val-Ala-Asp-7-amino-4-methylcoumarin

Specifications

Formula Weight 716.73g/mol
Synonym Ac-VDVAD-AMC; Caspase-2 Substrate (ICH-1) fluorogenic
Storage Requirements −30° to −10°C
Format Lyophilized Powder
Description White to Off-white
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66089LB0

 
alfa aesar™
J66089LB0
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66089MB

 
alfa aesar™
J66089MB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 716.73g/mol
Synonym Ac-VDVAD-AMC; Caspase-2 Substrate (ICH-1) fluorogenic
Storage Requirements −30° to −10°C
Format Lyophilized Powder
Description White to Off-white