Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Acetyl-Val-Ala-Asp-al

For cellular and molecular biology applications

$76.05 - $240.12

Specifications

Product Type N-Acetyl-Val-Ala-Asp-al
Molecular Weight 329.35
Storage Requirements -30° to -10°C
Sequence Acetyl-VAD-CHO
Format Lyophilized powder
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66696LB0

 
alfa aesar™
J66696LB0
5mg Each for $240.12

AAJ66696MCR

 
alfa aesar™
J66696MCR
1mg Each for $76.05
Description & Specifications

Specifications

Product Type N-Acetyl-Val-Ala-Asp-al
Molecular Weight 329.35
Storage Requirements -30° to -10°C
Sequence Acetyl-VAD-CHO
Format Lyophilized powder
Regulatory Status RUO
Purification Method HPLC